Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260858.2 | NP_001247787.1 | 117 | 102 | ||
GenBank mRNA | JU321726.1 | - | 174 | 102 | ||
JU473527.1 | - | 284 | 102 | |||
JV043991.1 | - | 119 | 102 | |||
JV651503.1 | - | 295 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |