Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU332724.1 | - | 296 | 73 | ||
JU479402.1 | - | 199 | 73 |