Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258129.1 | NP_001245058.1 | 968 | 100 | ||
XM_015121180.2 | XP_014976666.1 | 1335 | 100 | |||
GenBank mRNA | JU471196.1 | - | 968 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |