Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265711.1 | NP_001252640.1 | 3690 | 90 | ||
XM_028837093.1 | XP_028692926.1 | 6464 | 90 | |||
GenBank mRNA | JV565011.1 | - | 3690 | 90 | ||
JV712201.1 | - | 3691 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |