Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU337403.1 | - | 4142 | 74 | ||
JU337404.1 | - | 4228 | 74 | |||
JU478114.1 | - | 4203 | 74 | |||
JU478115.1 | - | 4181 | 74 |
Gene Symbol: TMEM243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194372.2 | NP_001181301.1 | 653 | 74 | ||
XM_015134078.2 | XP_014989564.1 | 524 | 74 | |||
XM_015134079.2 | XP_014989565.1 | 517 | 74 | |||
XM_015134080.2 | XP_014989566.1 | 572 | 74 | |||
GenBank mRNA | JV635628.1 | - | 451 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |