Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265650.1 | NP_001252579.1 | 382 | 90 | ||
XM_015117359.2 | XP_014972845.1 | 412 | 90 | |||
GenBank mRNA | JU326234.1 | - | 258 | 90 | ||
JV667335.1 | - | 382 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |