Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260739.1 | NP_001247668.1 | 249 | 62 | ||
GenBank mRNA | JU320685.1 | - | 235 | 62 | ||
JU472487.1 | - | 270 | 62 | |||
JV047478.1 | - | 249 | 62 | |||
JV654843.1 | - | 274 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |