Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114955.1 | NP_001108427.1 | 661 | 88 | ||
GenBank mRNA | FJ375751.1 | - | 661 | 88 |