Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088822.3 | XP_001088822.2 | 1172 | 56 | ||
XM_028836401.1 | XP_028692234.1 | 1250 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |