Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806209.3 | XP_002806255.2 | 304 | 97 | ||
GenBank mRNA | JU337313.1 | - | 76 | 97 | ||
JV048826.1 | - | 159 | 97 | |||
JV666670.1 | - | 162 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |