Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257765.1 | NP_001244694.1 | 1169 | 59 | ||
XM_015118307.2 | XP_014973793.2 | 1883 | 59 | |||
XM_015118308.2 | XP_014973794.2 | 1240 | 59 | |||
XM_015118309.2 | XP_014973795.2 | 1251 | 59 | |||
XM_028835628.1 | XP_028691461.1 | 1922 | 59 | |||
XM_028835629.1 | XP_028691462.1 | 2521 | 59 | |||
XM_028835630.1 | XP_028691463.1 | 1214 | 59 | |||
XM_028835631.1 | XP_028691464.1 | 2500 | 59 | |||
GenBank mRNA | JU333634.1 | - | 1169 | 59 | ||
JU474889.1 | - | 1164 | 59 | |||
JV576210.1 | - | 1171 | 59 | |||
JV677963.1 | - | 1176 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |