Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152448.2 | XP_015007934.1 | 893 | 150 | ||
XM_015152449.2 | XP_015007935.1 | 840 | 150 | |||
XM_028829779.1 | XP_028685612.1 | 900 | 150 | |||
XM_028829780.1 | XP_028685613.1 | 685 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |