Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261086.1 | NP_001248015.1 | 231 | 132 | ||
GenBank mRNA | JU332756.1 | - | 231 | 132 | ||
JU332757.1 | - | 231 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |