Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193501.2 | NP_001180430.1 | 578 | 87 | ||
GenBank mRNA | JU337410.1 | - | 273 | 87 | ||
JU494500.1 | - | 409 | 87 | |||
JV724632.1 | - | 393 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |