Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261767.1 | NP_001248696.1 | 322 | 88 | ||
GenBank mRNA | JU327774.1 | - | 285 | 88 | ||
JU474458.1 | - | 322 | 88 | |||
JV489642.1 | - | 332 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |