Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141795.2 | XP_014997281.1 | 726 | 133 | ||
XM_028850707.1 | XP_028706540.1 | 753 | 133 | |||
GenBank mRNA | JU321268.1 | - | 679 | 133 | ||
JU477234.1 | - | 597 | 133 | |||
JV048273.1 | - | 666 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |