Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261346.1 | NP_001248275.1 | 436 | 65 | ||
XM_015151147.2 | XP_015006633.1 | 321 | 65 | |||
XM_028828917.1 | XP_028684750.1 | 222 | 65 | |||
GenBank mRNA | JU321370.1 | - | 443 | 65 | ||
JV043382.1 | - | 436 | 65 | |||
JV048906.1 | - | 151 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |