Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193883.2 | NP_001180812.2 | 113 | 61 | ||
XM_015110595.2 | XP_014966081.2 | 303 | 61 | |||
XM_015110596.2 | XP_014966082.2 | 303 | 61 | |||
XM_015110597.2 | XP_014966083.1 | 195 | 61 | |||
GenBank mRNA | JU338818.1 | - | 112 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |