Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001110903.4 | XP_001110903.1 | 970 | 93 | ||
GenBank mRNA | JU481790.1 | - | 432 | 93 | ||
JV708683.1 | - | 454 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |