Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258149.2 | NP_001245078.1 | 107 | 145 | ||
GenBank mRNA | JU476814.1 | - | 107 | 145 | ||
JV625877.1 | - | 134 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |