Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266085.1 | NP_001253014.1 | 460 | 62 | ||
GenBank mRNA | JU327091.1 | - | 301 | 62 | ||
JU472813.1 | - | 460 | 62 | |||
JV048785.1 | - | 460 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |