Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143888.1 | XP_014999374.1 | 460 | 68 | ||
XM_015143889.2 | XP_014999375.1 | 443 | 68 | |||
GenBank mRNA | JV052451.1 | - | 399 | 68 | ||
JV642960.1 | - | 384 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |