Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193771.2 | NP_001180700.1 | 763 | 64 | ||
GenBank mRNA | JU320903.1 | - | 642 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |