Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142219.2 | XP_014997705.2 | 800 | 145 | ||
XM_028850897.1 | XP_028706730.1 | 731 | 145 | |||
XM_028850898.1 | XP_028706731.1 | 858 | 145 | |||
XM_028850899.1 | XP_028706732.1 | 789 | 145 | |||
XM_028850900.1 | XP_028706733.1 | 799 | 145 | |||
XM_028850901.1 | XP_028706734.1 | 799 | 145 | |||
GenBank mRNA | JU324318.1 | - | 632 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |