Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143527.2 | XP_014999013.1 | 272 | 149 | ||
GenBank mRNA | JU332587.1 | - | 158 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |