Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257903.2 | NP_001244832.1 | 603 | 124 | ||
XM_028843154.1 | XP_028698987.1 | 1450 | 124 | |||
GenBank mRNA | JU323034.1 | - | 779 | 124 | ||
JU323035.1 | - | 607 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |