Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105290.4 | XP_001105290.1 | 148 | 113 | ||
XM_028850346.1 | XP_028706179.1 | 145 | 113 | |||
XM_028850348.1 | XP_028706181.1 | 145 | 113 | |||
GenBank mRNA | JU335238.1 | - | 139 | 113 | ||
JU335239.1 | - | 139 | 113 | |||
JU472803.1 | - | 109 | 113 | |||
JU476206.1 | - | 109 | 113 | |||
JU476207.1 | - | 109 | 113 | |||
JU476208.1 | - | 109 | 113 | |||
JU476209.1 | - | 109 | 113 | |||
JV043974.1 | - | 109 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |