Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141422.2 | XP_014996908.1 | 1429 | 97 | ||
XM_015141423.2 | XP_014996909.1 | 1443 | 97 | |||
XM_028849922.1 | XP_028705755.1 | 383 | 97 | |||
GenBank mRNA | JU333128.1 | - | 179 | 97 | ||
JU472458.1 | - | 151 | 97 | |||
JV045363.1 | - | 300 | 97 | |||
JV643461.1 | - | 309 | 97 |