Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092385.4 | XP_001092385.2 | 1168 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |