Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115228.2 | XP_014970714.1 | 2008 | 75 | ||
XM_015115229.2 | XP_014970715.1 | 2005 | 75 | |||
XM_015115230.2 | XP_014970716.1 | 1981 | 75 | |||
XM_015115231.2 | XP_014970717.1 | 2008 | 75 | |||
XM_015115232.2 | XP_014970718.1 | 1978 | 75 | |||
XM_015115233.2 | XP_014970719.1 | 1855 | 75 | |||
XM_015115234.2 | XP_014970720.1 | 2008 | 75 | |||
XM_015115236.2 | XP_014970722.1 | 1828 | 75 | |||
XM_015115237.2 | XP_014970723.1 | 1855 | 75 | |||
XM_015115239.2 | XP_014970725.1 | 1981 | 75 | |||
XM_028833834.1 | XP_028689667.1 | 2008 | 75 | |||
GenBank mRNA | JU335540.1 | - | 1959 | 75 | ||
JU476467.1 | - | 1959 | 75 | |||
JV719156.1 | - | 1964 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |