Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082699.4 | XP_001082699.2 | 583 | 95 | ||
XM_028828184.1 | XP_028684017.1 | 7390 | 95 | |||
GenBank mRNA | JU333476.1 | - | 776 | 95 | ||
JU340584.1 | - | 477 | 95 | |||
JU474750.1 | - | 780 | 95 | |||
JV046478.1 | - | 770 | 95 | |||
JV709874.1 | - | 484 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |