Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261728.1 | NP_001248657.1 | 393 | 104 | ||
XM_015110573.2 | XP_014966059.1 | 507 | 104 | |||
XM_015110575.2 | XP_014966061.1 | 308 | 104 | |||
GenBank mRNA | JU472958.1 | - | 557 | 104 | ||
JU476842.1 | - | 419 | 104 | |||
JV044207.1 | - | 393 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |