Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032939.1 | NP_001028111.1 | 179 | 61 | ||
GenBank mRNA | AF366397.1 | - | 179 | 61 | ||
JU336136.1 | - | 296 | 61 | |||
JU476998.1 | - | 298 | 61 | |||
JV660075.1 | - | 320 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta Inventoried |