Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107218.4 | XP_001107218.2 | 1197 | 92 | ||
XM_015138869.2 | XP_014994355.1 | 1297 | 92 | |||
XM_028848641.1 | XP_028704474.1 | 1109 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |