Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36RN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832126.1 | XP_028687959.1 | 191 | 109 | ||
XM_028832127.1 | XP_028687960.1 | 304 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |