Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126450.2 | XP_014981936.1 | 684 | 81 | ||
XM_015126451.2 | XP_014981937.1 | 532 | 81 | |||
XM_028841542.1 | XP_028697375.1 | 457 | 81 | |||
XM_028841543.1 | XP_028697376.1 | 327 | 81 | |||
XM_028841544.1 | XP_028697377.1 | 341 | 81 | |||
GenBank mRNA | JV064396.1 | - | 275 | 81 | ||
JV472713.1 | - | 267 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |