Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260833.1 | NP_001247762.1 | 682 | 66 | ||
GenBank mRNA | JU333278.1 | - | 752 | 66 | ||
JU474589.1 | - | 703 | 66 | |||
JV046372.1 | - | 682 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |