Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266713.1 | NP_001253642.1 | 1207 | 102 | ||
XM_015118795.2 | XP_014974281.1 | 1415 | 102 | |||
XM_028843550.1 | XP_028699383.1 | 908 | 102 | |||
GenBank mRNA | JU322212.1 | - | 1306 | 102 | ||
JV489114.1 | - | 1207 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |