Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266409.1 | NP_001253338.1 | 411 | 106 | ||
XM_015120241.2 | XP_014975727.1 | 498 | 106 | |||
XM_015120242.2 | XP_014975728.1 | 495 | 106 | |||
XM_015120243.2 | XP_014975729.1 | 650 | 106 | |||
XM_015120244.2 | XP_014975730.1 | 473 | 106 | |||
XM_015120245.2 | XP_014975731.1 | 517 | 106 | |||
XM_028835495.1 | XP_028691328.1 | 502 | 106 | |||
XM_028835496.1 | XP_028691329.1 | 479 | 106 | |||
GenBank mRNA | JU333928.1 | - | 346 | 106 | ||
JV046722.1 | - | 411 | 106 | |||
JV711775.1 | - | 286 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |