Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYKPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260930.2 | NP_001247859.1 | 1296 | 73 | ||
XM_028849166.1 | XP_028704999.1 | 1451 | 73 | |||
XM_028849167.1 | XP_028705000.1 | 1403 | 73 | |||
XM_028849170.1 | XP_028705003.1 | 1172 | 73 | |||
GenBank mRNA | JU322930.1 | - | 1296 | 73 | ||
JU478189.1 | - | 1370 | 73 |