Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261078.1 | NP_001248007.1 | 486 | 65 | ||
XM_015134309.1 | XP_014989795.1 | 578 | 65 | |||
XM_015134321.1 | XP_014989807.1 | 622 | 65 | |||
XM_028843740.1 | XP_028699573.1 | 1243 | 65 | |||
XM_028843743.1 | XP_028699576.1 | 627 | 65 | |||
GenBank mRNA | JU333340.1 | - | 486 | 65 | ||
JU333341.1 | - | 486 | 65 | |||
JV043725.1 | - | 486 | 65 | |||
JV046425.1 | - | 486 | 65 | |||
JV644907.1 | - | 489 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |