Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261537.1 | NP_001248466.1 | 72 | 69 | ||
XM_015140028.2 | XP_014995514.1 | 247 | 69 | |||
GenBank mRNA | JU475253.1 | - | 72 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |