Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194056.1 | NP_001180985.1 | 660 | 66 | ||
GenBank mRNA | JV068471.1 | - | 614 | 66 | ||
JV635684.1 | - | 674 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |