Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261616.1 | NP_001248545.1 | 955 | 68 | ||
XM_015120886.2 | XP_014976372.1 | 1026 | 68 | |||
XM_015120887.2 | XP_014976373.1 | 849 | 68 | |||
GenBank mRNA | JU320676.1 | - | 961 | 68 | ||
JU473904.1 | - | 735 | 68 | |||
JV043739.1 | - | 955 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |