Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195759.1 | NP_001182688.1 | 749 | 72 | ||
GenBank mRNA | CB308133.1 | - | 190 | 72 | ||
JU334771.1 | - | 793 | 72 | |||
JU475813.1 | - | 801 | 72 | |||
JV045371.1 | - | 799 | 72 | |||
JV583494.1 | - | 782 | 72 | |||
JV600971.1 | - | 780 | 72 | |||
JV703544.1 | - | 795 | 72 |
Gene Symbol: LOC114675523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842742.1 | XP_028698575.1 | 762 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |