Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193776.3 | NP_001180705.2 | 383 | 108 | ||
GenBank mRNA | JV074903.1 | - | 113 | 108 | ||
JV664895.1 | - | 199 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |