Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257690.1 | NP_001244619.1 | 355 | 97 | ||
XM_015126452.2 | XP_014981938.1 | 433 | 97 | |||
GenBank mRNA | JU337183.1 | - | 355 | 97 | ||
JU477914.1 | - | 289 | 97 | |||
JV048755.1 | - | 154 | 97 | |||
JV598865.1 | - | 377 | 97 | |||
JV666054.1 | - | 371 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |