Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266195.1 | NP_001253124.1 | 909 | 86 | ||
XM_015138746.2 | XP_014994232.1 | 1251 | 86 | |||
GenBank mRNA | JU333117.1 | - | 972 | 86 | ||
JU333118.1 | - | 972 | 86 | |||
JU333119.1 | - | 972 | 86 | |||
JU333120.1 | - | 972 | 86 | |||
JU333121.1 | - | 972 | 86 | |||
JU333122.1 | - | 972 | 86 | |||
JU333123.1 | - | 972 | 86 | |||
JU333124.1 | - | 972 | 86 | |||
JU333125.1 | - | 972 | 86 | |||
JU474455.1 | - | 863 | 86 | |||
JV046281.1 | - | 909 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |