Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS37C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114045.2 | XP_014969531.2 | 1039 | 54 | ||
XM_015114047.2 | XP_014969533.2 | 541 | 54 | |||
XM_015114048.2 | XP_014969534.2 | 932 | 54 | |||
XM_028833375.1 | XP_028689208.1 | 983 | 54 | |||
XM_028833376.1 | XP_028689209.1 | 284 | 54 | |||
GenBank mRNA | JU323701.1 | - | 271 | 54 | ||
JV654910.1 | - | 295 | 54 |
Gene Symbol: LOC114675519 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842739.1 | XP_028698572.1 | 470 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |