Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257923.1 | NP_001244852.1 | 147 | 58 | ||
XM_028846419.1 | XP_028702252.1 | 140 | 58 | |||
GenBank mRNA | JU332259.1 | - | 147 | 58 | ||
JU473726.1 | - | 112 | 58 | |||
JV044736.1 | - | 136 | 58 | |||
JV565151.1 | - | 67 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |